Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-SRP68, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222307
Description
SRP68 Polyclonal specifically detects SRP68 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SRP68 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
signal recognition particle 68 kDa protein, signal recognition particle 68kD, signal recognition particle 68kDa | |
Rabbit | |
Affinity Purified | |
RUO | |
6730 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
NP_055045.2 | |
SRP68 | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Signal Recognition Particle 68kDa using the numbering given in entry NP_055045.2 (GeneID 6730). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only