Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SRPK2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130114

Catalog No. NBP130114


Add to cart

Description

Description

SRPK2 Polyclonal antibody specifically detects SRPK2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

SRPK2
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate
P78362
EC 2.7.11.1, FLJ36101, H_RG152G17.1a, H_RG152G17.1b, serine kinase SRPK2, Serine/arginine-rich protein-specific kinase 2, serine/arginine-rich splicing factor kinase 2, serine/threonine-protein kinase SRPK2, SFRS protein kinase 2SFRSK2, SR protein kinase 2, SR-protein-specific kinase 2, SRSF protein kinase 2, WUGSC:H_RG152G17.1a
Rabbit
IgG
0.1 ml
Protein Kinase
Primary
6733
Human
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
SRPK2
Antigen maps to a region between residue 425 and 475 of human SFRS protein kinase 2 using the numbering given in entry NP_872633.1
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Horse, Rabbit, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Little brown bat, and Northern white-cheeked gibbon.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only