Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-SRPR alpha, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP132421
Description
SRPR alpha Polyclonal antibody specifically detects SRPR alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SRPR alpha | |
0.37mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Affinity Purified | |
SRPR | |
Recombinant protein encompassing a sequence within the center region of human SRPR. The exact sequence is proprietary. | |
70 kDa | |
0.1 ml | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
Docking protein alpha, DP, DP-alpha, MGC17355, MGC3650, MGC9571, signal recognition particle receptor (docking protein), signal recognition particle receptor ('docking protein'), signal recognition particle receptor subunit alpha, Sralpha, SR-alpha, SRP-alpha | |
Rabbit | |
IgG | |
Immunogen affinity purified | |
RUO | |
Primary | |
6734 | |
Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only