Learn More
SRPRB, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Supplier:  Thermo Scientific PA130658
Catalog No. PIPA130658
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-30658 detects SRPR beta from canine samples.PA1-30658 has been successfully used in immunoprecipitation, Western blot applications.The PA1-30658 immunogen is: Synthetic peptide, corresponding to C terminal amino acids 246-265 of Dog SRPR beta.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SRPR beta
Polyclonal
Unconjugated
SRPRB
Synthetic peptide, corresponding to C terminal amino acids 246-265 of Dog SRPR beta.
250 μg
Primary
Canine
Antibody
IgG
Immunoprecipitation, Western Blot
1mg/mL
SRPRB
Sheep
Ammonium sulfate precipitation
RUO
610826
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Recommendations
Recommendations