Learn More
anti-SSEA-1, Alexa Fluor 700, Clone: MC-480, R&D Systems™
Manufacturer:  R&D Systems FAB2155N025
 View more versions of this product
Catalog No. FAB2155N025
Add to cart
Specifications
Specifications
SSEA-1
MC-480
Flow Cytometry 5 uL/10^6 cells
Affinity Purified
FUT4
F9 mouse teratocarcinoma stem cells
IgM-specific Affinity-purified from hybridoma culture supernatant
RUO
Primary
2526
Human, Mouse
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA1, SSEA-1, stage-specific embryonic antigen 1
Mouse
IgM
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human and mouse SSEA-1.