Learn More
anti-SSEA-4, PerCP, Clone: MC-813-70, R&D Systems™
Manufacturer:  R&D Systems FAB1435C025
 View more versions of this product
Catalog No. FAB1435C025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
SSEA-4
MC-813-70
Supplied in a saline solution containing BSA and Sodium Azide.
Murine
IgG3
25 Tests
Primary
Recognizes a carbohydrate epitope of SSEA-4.
Flow Cytometry
PerCP
Purified
2120Ep human embryonal carcinoma cell line
Protein A or G purified from hybridoma culture supernatant
RUO
Monoclonal
Human, Murine