Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-SSEA-5; Clone: 8e11/SSEA-5; BD
Supplier: BD Biosciences 563752
Description
Host Species: Mouse
Clone: 8e11/SSEA-5
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human Undifferentiated Embryonic Stem Cells
Flow cytometry, Immunofluorescence, Bioimaging, Cell Depletion, Western blotting
Specifications
SSEA-5 | |
8e11/SSEA-5 | |
Unconjugated | |
Human Undifferentiated Embryonic Stem Cells | |
0.1 mg | |
Primary |
Monoclonal | |
0.5mg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction