Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSR4, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127913
Description
TRAP proteins are part of a complex whose function is to bind calcium to the ER membrane and thereby regulate the retention of ER resident proteins. SSR4. also called TRAPD. is assumed to be involved in protein secretion. It is located in the Xq28 region.
PA1-27913 detects SSR4 from human, mouse, and rat samples.PA1-27913 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Translocon-associated protein subunit delta.
Specifications
| SSR4 | |
| Polyclonal | |
| SSR4 | |
| TRAPD | |
| Chicken | |
| Antigen affinity chromatography | |
| RUO | |
| 6748 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| P51571, Q62186, Q07984 | |
| SSR4 | |
| Synthetic peptide corresponding to human Translocon-associated protein subunit delta. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction