Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSRP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112698

Catalog No. 01-674-837


Add to cart

Description

Description

SPT16 and SSRP1 are found in all eurkaryotic cells and exists as part of an essential heterodimer complex. Together, SPT 16 and SSRP1 are a highly abundant nuclear complex that in mammals has been called FACT, or Facilitates Chromatin Transcription. FACT enhances the processivity of RNA polymerase II through nucleosomes. Evidence also points to a role for FACT in replication, possibly by enhancing DNA polymerase through nucleosomes as well.
Specifications

Specifications

SSRP1
Polyclonal
Unconjugated
SSRP1
Liquid
SSRP1
Full-length recombit SSRP1 protein tagged with histidine.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse
Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.08% sodium azide
Q08945, Q08943
Structure specific recognition protein 1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
20833, 6749
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit