Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ST2/IL-1 R4, Polyclonal, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233319

Catalog No. NBP233319

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ST2/IL-1 R4
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
suppression of tumorigenicity 2
Sheep
IgG
0.1 mg
Innate Immunity
Primary
9173
Reconstitute with 0.1 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Western Blot, Blocking Assay
Western Blot 1:1000-1:2000, Blocking/Neutralizing
Affinity Purified
IL1RL1
E. coli-derived recombinant human ST2/IL-1 R4
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Detects human ST2/IL-1 R4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-18 RI, and rhST-7 is observed.
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only