Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST7 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523995
Description
This antibody is predicted to react with bovine, chicken, mouse, porcine, non-human primate, rat, rabbit and zebrafish based on sequence homology.
The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described.Specifications
ST7 | |
Polyclonal | |
ST7 | |
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7, family with sequence similarity 4, subfamily A, member 1, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7982 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q9NRC1 | |
ST7 | |
KLH conjugated synthetic peptide between 169-197 amino acids from the central region of human ST7 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction