Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179292
Description
ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 in Human samples. It is validated for Western Blot.Specifications
ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Alpha-2,8-sialyltransferase 8D, CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase, CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase, EC 2.4.99, EC 2.4.99.-, Polysialyltransferase-1, PST1MGC61459, PSTMGC34450, sialyltransferase 8 (alpha-2, 8-polysialytransferase) D, Sialyltransferase 8D, Sialytransferase St8Sia IV, SIAT8D, SIAT8-D, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4, ST8SiaIV, ST8SIA-IV | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Equine: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Guinea pig: 85%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_005659 | |
ST8SIA4 | |
Synthetic peptide directed towards the middle region of human ST8SIA4The immunogen for this antibody is ST8SIA4. Peptide sequence DVGTKSDFITMNPSVVQRAFGGFRNESDREKFVHRLSMLNDSVLWIPAFM. | |
100 μL | |
Neuroscience | |
7903 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction