Learn More
STAC Mouse anti-Human, Clone: 2C5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA520035
Catalog No. PIMA520035
Add to cart
Description
Description
Peptide Sequence: LAQRTKKGSS GSGSDSPHRT STSDLVEVPE EANGPGGGYD LRKRSNSVFT YPENGTDDFR DPAKNINHQG SLSKDPLQMN TYVALYKFVP QENEDLEMRP GDIITLLEDS
Probably involved in a neuron-specific signal transduction.
Specifications
Specifications
STAC
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q99469
STAC1, SH3 and cysteine-rich domain-containing protein, SRC homology 3 and cysteine-rich domain-containing protein, src homology three (SH3) and cysteine rich domain
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6769
ELISA, Western Blot
2C5
Unconjugated
STAC
Liquid
STAC
STAC (AAH20221, 209 a.a. ∽ 318 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human