Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-STAG2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130472

Catalog No. NBP130472


Add to cart

Description

Description

STAG2 Polyclonal antibody specifically detects STAG2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

STAG2
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Q8N3U4
bA517O1.1, cohesin subunit SA-2, DKFZp686P168, SA-2, SA2FLJ25871, SCC3 homolog 2, stromal antigen 2DKFZp781H1753
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
STAG2
The immunogen for this product maps to a region between residue 1 and 50 of human stromal antigen 2 using the numbering given in entry NP_006594.3 (GeneID 10735).
141 kDa
0.1 ml
DNA Repair
Primary
10735
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only