Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAMBPL1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525246
Description
STAMBPL1 is zinc metalloprotease that specifically cleaves ′Lys-63′-linked polyubiquitin chains. Does not cleave ′Lys-48′-linked polyubiquitin chains.Specifications
STAMBPL1 | |
Polyclonal | |
STAMBPL1 | |
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2, AMSH-like protease, associated molecule with the SH3 domain of STAM (AMSH) - Family Protein, associated molecule with the SH3 domain of STAM (AMSH) like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57559, 76630 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q96FJ0, Q76N33 | |
STAMBPL1 | |
KLH conjugated synthetic peptide between 135-163 amino acids from the central region of human STAMBPL1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction