Learn More
MilliporeSigma™ anti-Stargazin, Polyclonal, ™ (Chemicon™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB9876/DEL
Catalog No. 50-174-298
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
L-type calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium chan
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
Stargazin
Unconjugated
Rabbit
100μg
Neuroscience
Polyclonal
0.145mg/mL
Affinity Purified
GST fusion protein from rat stargazin, cytoplasmic domain.
RUO
Primary
Murine, Rat