Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stargazin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061089
Description
Stargazin Polyclonal specifically detects Stargazin in Mouse, Rat samples. It is validated for Western Blot.Specifications
Stargazin | |
Polyclonal | |
Western Blot 1:1000 | |
Q71RJ2 | |
CACNG2 | |
Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. Accession # O88602 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼36k stargazin protein. Immunolabeling of the stargazin band is blocked by preadsorption with the peptide used as antigen. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
calcium channel, voltage-dependent, gamma subunit 2, FLJ41437, MGC138502, MGC138504, Neuronal voltage-gated calcium channel gamma-2 subunit, stargazin, voltage-dependent calcium channel gamma-2 subunit | |
Rabbit | |
36 kDa | |
0.1 mL | |
Neuroscience | |
10369 | |
Mouse, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction