Learn More
anti-Stargazin, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP146404
Catalog No. NBP146404
Add to cart
Description
Description
Stargazin Polyclonal antibody specifically detects Stargazin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Stargazin
LYOPH
Western Blot 1: 300-1: 2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Antisera
CACNG2
A synthetic peptide from the internal region of human CACNG2 conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat, mouse and zebrafish.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specific for CACNG2.
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Whole antisera with No Preservative
calcium channel, voltage-dependent, gamma subunit 2, FLJ41437, MGC138502, MGC138504, Neuronal voltage-gated calcium channel gamma-2 subunit, stargazin, voltage-dependent calcium channel gamma-2 subunit
Goat
IgG
0.1 ml
Neuroscience
Primary
10369
Reconstitute with 0.1 ml sterilized water to desired concentration.