Learn More
STAT2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06502
Catalog No. 06-502-MI
Add to cart
Description
Description
Anti-STAT2 Antibody, CT is a Rabbit Polyclonal Antibody for detection of STAT2 also known as Signal Transducer and Activator of Transcription 2 and has been tested in EMSA, immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
STAT2
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_005419
Electromobility Shift Assay, Immunocytochemistry, Immunoprecipitation, Western Blot
0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
STAT2; ISGF-3; P113; p113; STAT113; MGC59816
GST fusion protein containing amino acids 671-806 of human STAT2
Protein A chromatography
−20°C for 24 months
Polyclonal
Human, Murine