Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat2 Mouse anti-Human, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610188

 View more versions of this product

Catalog No. BDB610188


Add to cart

Description

Description

The Stat proteins function as both cytoplasmic signal transducers and activators of transcription. Stat91/84 (the two proteins are the result of alternate splicing-Stat91 has an additional 38 C-terminal amino acids) and Stat113 were the first identified members of this protein family. These three polypeptides contain both SH2 and SH3 domains and have also been described as members of the ISGF3 (interferon-stimulated gene factor 3) complex. With the discovery of additional members of the Stat family (Stats3 & 4), the nomenclature has been revised to indicate the Stat family members in the order of their discovery. Stat 91, 84, and 113 have become Stat1α, Stat1β, and Stat2, respectively. In response to IFN-α treatment, Stat1α, Stat1β, and Stat2 become tyrosine-phosphorylated and migrate to the nucleus where they join a 48kDa DNA binding protein and subsequently direct the transcription at IFN-α responsive elements. The genes encoding Stat2 and Stat1 are closely related. An extended acidic C-terminal region exclusive to Stat2 implicates a potential role in gene activation. Stat2 phosphorylation can occur independently of Stat1α, but phosphorylated Stat2 does appear to be dependent on its interaction with Stat1α for efficient nuclear localization and/or stability.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Stat2
22
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.