Learn More
anti-Stat3, PerCP-Cy 5.5; Clone: M59-50; BD
Manufacturer:  BD Biosciences 564133
Catalog No. BDB564133
Add to cart
Description
Description
Applications:
Flow cytometry
Specifications
Specifications
Stat3
M59-50
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
IgG1, lambda
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Monoclonal
Flow Cytometry
PerCP-Cyanine5.5
Affinity purified
Human Stat3 Recombinant Protein
RUO
Primary
Human, Murine, Rat