Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat4 Mouse, Unlabeled, Clone: 8, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610926

Catalog No. BDB610926


Add to cart

Description

Description

The Stat proteins function as both cytoplasmic signal transducers and activators of transcription. Stat91/84 (the two proteins are the result of alternate splicing-Stat91 has an additional 38 C-terminal amino acids) and Stat113 were the first identified members of this protein family. With the discovery of additional members of the Stat family (Stat 3 & 4), the nomenclature has been revised to reflect the order of their discovery. Stat 91, 84, and 113 have become Stat1α, Stat1β, and Stat2, respectively. Stat4 has been reported to be 52% identical to Stat1 and 47% identical to Stat3. Functionally, Stat4 is similar to the other Stat proteins in that it requires tyrosine phosphorylation in order to stimulate its IFN-γ activated site (GAS) binding activity. Expression of Stat4 appears to be limited to thymus, spleen, and testis. Its expression in several myeloid cell lines is differentially regulated during cellular differentiation. Co-transfection studies using JAK1 or JAK2 expression constructs indicate that Stat4 is a substrate for these kinases.

Immunofluorescence, Western Blotting

Specifications

Specifications

Stat4
8
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Signal Transducers and Activators of Transcription-4
Affinity Purified
Human Stat4 aa. 136-258
50μg
Cell Biology
Primary
Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit