Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat5 (pY694) Rabbit anti-Human, Unlabeled, Polyclonal, BD

Rabbit Polyclonal Antibody

Manufacturer:  BD Biosciences 611818

 View more versions of this product

Catalog No. BDB611818


Add to cart

Description

Description

Mammary gland factor (MGF) has been shown to play a critical role in the lactogenic hormone response and has been well characterized in mammary epithelial cells. The SH3 domain of MGF shows extensive homology with the SH2 domains of Stat1 and Stat2. This homology leads to speculation that MGF may have a broader physiological role than originally anticipated and has led to its classification as a member of the Stat family. Stat5 (MGF) mRNA expression is highest in mammary gland tissue. Lower levels are found in ovary, thymus, spleen, kidney, lung, muscle, and the adrenal gland. The peptide hormone, prolactin, binds to the prolactin receptor (PRLR) to initiate the lactogenic response. There are at least three forms of PRLR; however, only the long form is able to activate the 92kDa Stat5 protein by inducing phosphorylation at Tyr694. Once phosphorylated, Stat5 becomes an essential transcription factor which binds to the β-casein gene promoter. Stat5 activity is tightly regulated throughout gestation,lactation, and post-lactation. Treatment of activated Stat5 with a protein tyrosine phosphatase results in the loss of DNA binding activity. The presence of an SH2 domain within Stat5 suggests that it may directly interact with protein tyrosine kinases (PTKs) such as JAK2.

Host Species: Rabbit
Clone: xSpecies Reactivity [for Features Main]: Human

Immunofluorescence, Western Blotting

Specifications

Specifications

Stat5 (pY694)
POLYCLONAL
Unconjugated
Rabbit
50μg
Cell Biology
Polyclonal
Immunofluorescence, Western Blot
250μg/mL
Purified
Ig
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.