Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-STAT5a (p Tyr694), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177851

Catalog No. NBP177851


Add to cart

Description

Description

STAT5A p Tyr694 Polyclonal antibody specifically detects STAT5A p Tyr694 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

STAT5a (p Tyr694)
1.1 mg/ml
Western Blot 1:500-1:2000, ELISA 1:25000, Immunohistochemistry 20-40 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
P42230
MGF, signal transducer and activator of transcription 5A, STAT5
Rabbit
IgG
0.1 mg
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary
6776
Human, Mouse, Rat
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
STAT5A
This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y694 of mouse STAT5a protein.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific for mouse Stat5a protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only