Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Stat6, PE-CF594; Clone: 23/Stat6; BD
Supplier: BD Biosciences 564148
Description
Applications:
Flow cytometry

Specifications
Stat6 | |
Monoclonal | |
PE-CF594 | |
Human Stat6 C-terminal Recombinant Protein | |
RUO | |
Bovine, Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
23/Stat6 | |
Mouse | |
Affinity Purified | |
Primary | |
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction