Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STC2, Polyclonal, Invitrogen™

Catalog No. PIPA127710
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127710 50 μg
1 options

Catalog No. PIPA127710

Supplier: Thermo Scientific PA127710

Chicken Polyclonal Antibody

Has an anti-hypocalcemic action on calcium and phosphate homeostasis. This protein is a secreted. homodimeric glycoprotein that is expressed in a wide variety of tissues and may have autocrine or paracrine functions. The encoded protein has 10 of its 15 c

PA1-27710 detects STC2 from human samples.PA1-27710 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Stanniocalcin-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STC2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene STC2
Gene Accession No. O76061
Gene Alias STC-2, STCRP
Gene Symbols STC2
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Stanniocalcin-2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.