Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STEAP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520406
Description
A suggested positive control is rat liver tissue lysate. PA5-20406 can be used with blocking peptide PEP-0523.
STEAP3 Polyclonal antibody specifically detects STEAP3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
STEAP3 | |
Polyclonal | |
Unconjugated | |
STEAP3 | |
1010001D01Rik; AHMIO2; dudlin 2; dudlin-2; dudulin 2; dudulin-2; hpHyde; hTSAP6; Metalloreductase STEAP3; pHyde; Protein nm1054; six-transmembrane epithelial antigen of prostate 3; STEAP family member 3; STEAP family member 3, metalloreductase; STEAP3; STEAP3 metalloreductase; STMP3; TSAP6; tumor suppressor pHyde; tumor suppressor-activated pathway protein 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170824, 55240, 68428 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5RKL5, Q658P3, Q8CI59 | |
STEAP3 | |
A 15 amino acid peptide from near the amino terminus of human STEAP3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction