Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-STIM1, DyLight 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11060547G

Catalog No. NB11060547G


Add to cart

Description

Description

STIM1 Polyclonal specifically detects STIM1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications

Specifications

STIM1
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
STIM1
STIM1 antibody was made to a synthetic peptide from the C-terminal region (within residues 600-685) of the human STIM1 protein. [UniProt# Q13586]
77 kDa
0.1 ml
Cancer, Cell Cycle and Replication, Plasma Membrane Markers, Signal Transduction
Primary
6786
Human, Mouse, Rat, Bovine, Canine
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated
Q13586
GOKD11S4896E, stromal interaction molecule 1
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, mouse, rat, bovine and canine. Immunogen has 93% homology to porcine and 79% homology to Xenopus.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only