Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STIM1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523623

Catalog No. PIPA523623


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

This gene encodes a type 1 transmembrane protein that mediates Ca2+ influx after depletion of intracellular Ca2+ stores by gating of store-operated Ca2+ influx channels (SOCs). It is one of several genes located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocrotical carcinoma, and lung, ovarian, and breast cancer. This gene may play a role in maligcies and disease that involve this region, as well as early hematopoiesis, by mediating attachment to stromal cells. This gene is oriented in a head-to-tail configuration with the ribonucleotide reductase 1 gene (RRM1), with the 3 ft. end of this gene situated 1.6 kb from the 5 ft. end of the RRM1 gene.
Specifications

Specifications

STIM1
Polyclonal
PBS with 0.09% sodium azide
Q13586
D11S4896E, GOK, STIM1L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6786
Immunohistochemistry, Western Blot
Unconjugated
STIM1
Liquid
STIM1
KLH conjugated synthetic peptide between 509-538 amino acids from the C-terminal region of human STIM1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit