Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-STK11IP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130109

Catalog No. NBP130109


Add to cart

Description

Description

STK11IP Polyclonal antibody specifically detects STK11IP in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

STK11IP
1.0 mg/mL
Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Affinity Purified
STK11IP
maps to a region between residue 400 and 450 of human serine/threonine kinase 11 interacting protein using the numbering given in entry NP_443134.2
Immunogen affinity purified
RUO
Primary
114790
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
KIAA1898LKB1 interacting protein, LIP1serine/threonine kinase 11-interacting protein, LKB1-interacting protein 1, LKB1IPserine/threonine-protein kinase 11-interacting protein, serine/threonine kinase 11 interacting protein, STK11IP1STK11 interacting protein
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only