Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK33 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523081

Catalog No. PIPA523081


Add to cart

Description

Description

Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.

Serine Threonine Kinase 33 is normally expressed in embryonic and germ-line cells, which ultimately differentiate into testis, lung, heart, brain and spinal cord. It is critical in both spermatogenesis and organ ontogenesis, and is thus a critical developmental protein. However, in many maligt tissues, STK33 is abnormally expressed, and its appearance in these tissues usually indicates an increase in the cancer cell viability. In cancer cells, STK33 suppresses mitochondrial apoptosis, which precludes normal processes to identify and eliminate cancerous cells.
Specifications

Specifications

STK33
Polyclonal
Unconjugated
STK33
Liquid
STK33
A synthetic peptide made to an internal region of the human Stk33 (within residues 1-150).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.65 mg/mL
PBS with 30% glycerol and 0.05% sodium azide
Q9BYT3, Q924X7
serine/threonine-protein kinase 33
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
117229, 65975
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit