Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK35 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514082

Catalog No. PIPA514082


Add to cart

Description

Description

STK35 can interact with PDLIM1/CLP-36 in the cytoplasm and localize to actin stress fibers. The encoded kinase may be a regulator of actin stress fibers in nonmuscle cells. This protein has also been found in the nucleus.
Specifications

Specifications

STK35
Polyclonal
PBS with 0.09% sodium azide
Q8TDR2, Q80ZW0
Serine/threonine kinase 35, CLP-36 interacting kinase, Clik1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140901, 67333
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
STK35
Liquid
STK35
KLH conjugated synthetic peptide between 374-404 amino acids from the central region of human STK35
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit