Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK38 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513831

Catalog No. PIPA513831


Add to cart

Description

Description

Stk38,also known as NUCLEAR DBF2-RELATED PROTEIN, contains 1 protein kinase domain that interacts with mob1 and mob2.The homodimeric s100b binds to two molecules of stk38. It is ubiquitously expressed with highest levels observed in peripheral blood leukocytes. Stk38 activated by binding of s100b which releases autoinhibitory n-lobe interactions, enabling atp to bind and the autophosphorylation of ser-281. thr-444 then undergoes calcium- dependent phosphorylation by an upstream kinase. Interactions between phosphorylated thr-444 and the n-lobe promote additional structural changes that complete the activation of the kinase. Autoinhibition is also released by the binding of mob1/mobkl1a and mob2/hcca2 to the n-terminal of stk38.
Specifications

Specifications

STK38
Polyclonal
PBS with 0.09% sodium azide
Q15208
Serine/threonine-protein kinase 38; NDR1 protein kinase; Nuclear Dbf2-related kinase 1; STK38; NDR1;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11329
Western Blot
Unconjugated
STK38
Liquid
STK38
KLH conjugated synthetic peptide between 410-439 amino acids from the C-terminal region of human STK38
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit