Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK38L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514803

Catalog No. PIPA514803

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal. Northern blot analysis revealed expression of a 3.9-kb transcript in all tissues tested, with the possible exception of adult brain. Highest expression was in peripheral blood leukocytes. Immunoblot and immunofluorescence microscopy demonstrated predomitly nuclear expression of a 55-kD protein.
Specifications

Specifications

STK38L
Polyclonal
PBS with 0.09% sodium azide
Q9Y2H1
Serine/threonine-protein kinase 38-like; NDR2 protein kinase; Nuclear Dbf2-related kinase 2; STK38L; KIAA0965; NDR2;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23012
Western Blot
Unconjugated
STK38L
Liquid
STK38L
KLH conjugated synthetic peptide between 408-437 amino acids from the C-terminal region of human STK38L
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit