Learn More
anti-STK40, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP133391
Catalog No. NBP133391
Add to cart
Description
Description
STK40 Polyclonal specifically detects STK40 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
STK40
0.97mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
STK40
Recombinant protein encompassing a sequence within the center region of human serine/threonine kinase 40. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Not yet tested in any other species.
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
2310004N11Rik, EC 2.7.11, EC 2.7.11.1, MGC4796, Ser/Thr-like kinase, serine/threonine kinase 40, serine/threonine-protein kinase 40, SGK495, SgK495SHIKRP11-268J15.4, SINK-homologous serine/threonine-protein kinase, Sugen kinase 495
Rabbit
IgG
0.1 ml
Protein Kinase
Primary
83931
Human