Learn More
Streptococcus pyogenes Group A Goat anti-Bacteria, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA127089
Catalog No. PA127089
Add to cart
Description
Description
PA1-27089 detects Streptococcus pyogenes Group A Carbohydrate from bacteria samples. PA1-27089 has been successfully used in ELISA, immunohistochemistry (frozen), immunoprecipitation, dot blot and Western blot procedures. The PA1-27089 immunogen is killed Streptococcus pyogenes vaccine Strain J17A4.
Specifications
Specifications
Streptococcus pyogenes Group A
Polyclonal
Unconjugated
Liquid
Goat
IgG
500 μg
4° C
Primary
Bacteria
Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
3.1 mg/mL
0.01M PBS with 0.1% sodium azide; pH 7.2
S. pyogenes
Killed Streptococcus pyogenes vaccine Strain J17A4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal