Learn More
Streptococcus agalactiae Mouse anti-Bacteria, Clone: Strep12-448.4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110873
Catalog No. 01-671-882
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10873 detects Streptococcus agalactiae from Strep agalactiae samples. MA1-10873 has been successfully used in ELISA applications. The MA1-10873 immunogen is UV-inactivated Strep agalactiae cells isolated from mastitic milk.
Once thought to infect only cows, where it produces mastitis, Streptococcus agalactiae is now known to be able to cause serious disease, bacteremia and meningitis, in immunocompromised individuals and in neonates. Infections in the newborn are the most common and are extremely serious. They are of two types. Early onset of infection, usually occuring within 5 days of birth, is manifested by bacteremia and pneumonia. This infection is contracted vertically, as the baby passes through the birth canal. The other type of infection is a meningitis that occurs between the 10th and 60th days of life. The origin of this infection is not known, but it is not believed to be the mother.
Specifications
Specifications
Streptococcus agalactiae
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
S. agalactiae
UV-inactivated Strep agalactiae cells isolated from mastitic milk.
Purified
RUO
Antibody
Monoclonal
ELISA
Strep12-448.4
Unconjugated
Liquid
Mouse
IgM
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Bacteria