Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus Group A Mouse anti-Bacteria, Clone: D30I, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA17278
Description
The MA1-7278 antibody reacts with Streptococcus Group A in bacterial and infected tissue samples. MA1-7278 has been successfully used in ELISA applications. The MA1-7278 antibody was raised against Strep. Group A antigen.
The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.Specifications
Streptococcus Group A | |
Monoclonal | |
0.1 mg/mL | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2a |
ELISA | |
D30I | |
Unconjugated | |
Strep. Group A antigen. | |
100 μg | |
Primary | |
4° C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title