Learn More
Streptococcus pneumoniae Mouse anti-Bacteria, Clone: Q51F, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518263
Catalog No. PIMA518263
Add to cart
Description
Description
This antibody is reactive with the S. pneumoniae common C-polysaccharide (backbone epitope). Purity is >90%.
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
Specifications
Specifications
Streptococcus pneumoniae
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
S. pneumoniae; Pneumococcus
Strep pneumoniae native protein
Protein A
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence
Q51F
Unconjugated
Liquid
Mouse
IgG3
100 μg
4° C
Primary
Bacteria