Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Striatin Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

Supplier:  MilliporeSigma AB5779

Catalog No. AB5779MI


Only null left
Add to Cart

Description

Description

Anti-Striatin Antibody detects level of Striatin and has been published and validated for use in immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Striatin
Polyclonal
Rabbit serum. Liquid containing 40% glycerol and 0.02% sodium azide.
Rabbit
100 μL
NM_003162.2
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
STRN; MGC125642; SG2NA; Striatin.
Synthetic Striatin peptide.
Primary
Human, Mouse, Rat
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only