Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STX10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526924

Catalog No. PIPA526924


Add to cart

Specifications

Specifications

STX10
Polyclonal
PBS with 0.09% sodium azide
O60499
SYN10, hsyn10, syntaxin-10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8677
Western Blot
Unconjugated
STX10
Liquid
STX10
KLH conjugated synthetic peptide between 60-89 amino acids from the N-terminal region of human STX10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit