Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512425

Catalog No. PIPA512425

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

STX3 is otentially involved in docking of synaptic vesicles at presynaptic active zones.
Specifications

Specifications

Syntaxin 3
Polyclonal
PBS with 0.09% sodium azide
Q13277
Syntaxin-3; STX3; STX3A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6809
Immunohistochemistry, Western Blot
Unconjugated
STX3
Liquid
STX3
KLH conjugated synthetic peptide between 178-205 amino acids from the central region of human STX3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit