Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

syntaxin 4, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant STX4A.

Manufacturer:  Abnova Corporation H00006810A01

Catalog No. 89-004-328


Add to cart

Description

Description

Sequence: DKERVALVVHPGTARLGSPDEEFFHKVRTIRQTIVKLGNKVQELEKQQVTILATPLPEESMKQELQNLRDEIKQLGREIRLQLKAIEPQKEEADENYNSVN
Specifications

Specifications

syntaxin 4
Unconjugated
50% glycerol
NM_004604
STX4A|p35-2
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6810
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant STX4A.
STX4
Serum
STX4
STX4A (NP_004595, 19 a.a. ~ 119 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit