anti-Su(var)3-9, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 4034000201ML

 View more versions of this product

Catalog No. 40-340-00201M

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Disclaimers:

For Research Use Only


    Specifications

    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Host Species Rabbit
    Applications Chromatin Immunoprecipitation,ELISA,Western Blotting
    Quantity 0.1mL
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Test Specificity Drosophila .
    Concentration 1mg/mL
    Conjugate Unlabeled
    Cross Reactivity Drosophila
    Format Affinity Purified
    Immunogen In vivo generated recombinant protein fragment.
    Regulatory Status RUO
    Antigen Su(var)3-9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit