Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Substance P, Polyclonal, Novus Biologicals™

Guinea Pig Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234223

Catalog No. NBP234223


Add to cart

Description

Description

Substance P Polyclonal antibody specifically detects Substance P in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Substance P
1 mg/mL
Immunohistochemistry 15 ug/ml
Affinity Purified
TAC1
NH2-CRPKPQQFFGLM-CONH2 Corresponding to residues 1-11 of rat Substance P
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Immunohistochemistry
Unconjugated
PBS with 0.05% Sodium Azide
neurokinin 1, neurokinin 2, neurokinin A, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma), neuropeptide gamma, neuropeptide K, NK2, NKA, NKNAneurokinin alpha, NPK, PPT, protachykinin-1, substance K, substance P, TAC2neuromedin L, tachykinin 2, tachykinin, precursor 1, tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2
Guinea Pig
IgG
0.1 ml
Growth and Development, Neuroendocrinology, Neuroscience
Primary
6863
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only