Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULF2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112864

Catalog No. 01-675-844


Add to cart

Description

Description

A suggested positive control for this product is human brain lysate.

Sulf-2 exhibits arylsulfatase activity and highly specific endoglucosamine-6-sulfatase activity. It can remove sulfate from the C-6 position of glucosamine within specific subregions of intact heparin. Mammalian Sulfs are endoproteolytically processed and secreted into the extracellular space of transfected cells, where they exhibit both arylsulfatase activity and highly specific endoglucosamine-6-sulfatase activity against intact heparin. Though the molecular weight of full length human Sulf2 is 100455 Da (870 amino acids), the endogenous Sulf2 is found by western blot to be present at molecular weights of 24.4, 33.7 and 51–60 kDa. Higher molecular weight species may be found depending on tissue and/or cell type.
Specifications

Specifications

Sulf2
Polyclonal
PBS with 0.08% sodium azide
Q8IWU5
RP5-1049G16.1, DKFZp313E091, FLJ44488, FLJ90554, HSULF-2, KIAA1247, MGC126411
Rabbit
IgG
50 μg
-80° C, Avoid Freeze/Thaw Cycles
Primary
55959
Western Blot
Unconjugated
SULF2
Liquid
SULF2
Synthetic peptide corresponding to human Sulf2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit