Learn More
Sulfaguanidine Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175427
Catalog No. PIPA175427
Add to cart
Description
Description
PA1-75427 specifically detects Sulfaguanidine. PA1-75427 has been successfully used in ELISA applications. The PA1-75427 immunogen is Sulguanidine-BSA.
Sulfaguanidine is a guanidine derivative of sulfanilamide used in veterinary medicine. Sulfaguanidine is poorly absorbed from the gut but is well suited for the treatment of bacillary dysentery and other enteric infections. [National Center for Biotechnology Information. PubChem Compound Database; CID=5324]
Specifications
Specifications
Sulfaguanidine
Polyclonal
PBS with 0.09% sodium azide
Sulphaguanidine; Sulfaguanidin; Guanicil; Sulfaguine; C7H10N4O2S
Sulguanidine-BSA
Purified
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Liquid
Sheep
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical