Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Sulfamidase/SGSH, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132061

Catalog No. NBP132061


Add to cart

Description

Description

Sulfamidase/SGSH Polyclonal antibody specifically detects Sulfamidase/SGSH in Human samples. It is validated for Western Blot.
Specifications

Specifications

Sulfamidase/SGSH
2.0mg/mL
Western Blot 1:500-1:3000
Purified
SGSH
Recombinant protein encompassing a sequence within the center region of human SGSH. The exact sequence is proprietary.
57 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identitiy: Xenopus tropicalis (87%). Not yet tested in any other species.
Western Blot
Unconjugated
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
heparan sulfate sulfatase, HSSN-sulphoglucosamine sulphohydrolase, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMDEC 3.10.1.1, Sulfoglucosamine sulfamidase, Sulphamidase
Rabbit
IgG
Protein A purified
RUO
Primary
6448
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only