Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT1C3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524548

Catalog No. PIPA524548


Add to cart

Description

Description

Has low sulphotransferase activity towards various substrates with alcohol groups (in vitro). May catalyze the sulfate conjugation of xenobiotic compounds and endogenous substrates.
Specifications

Specifications

SULT1C3
Polyclonal
PBS with 0.09% sodium azide
Q6IMI6
ST1C3, sulfotransferase 1C3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
442038
Western Blot
Unconjugated
SULT1C3
Liquid
SULT1C3
KLH conjugated synthetic peptide between 264-292 amino acids from the C-terminal region of human SULT1C3
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit