Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMF2 Monoclonal Antibody (330312), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524058
Description
No cross-reactivity with recombinant human SUMF1 or recombinant mouse SUMF2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
SUMF2 Monoclonal antibody specifically detects SUMF2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
SUMF2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8NBJ7 | |
SUMF2 | |
Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunoprecipitation, Western Blot | |
330312 | |
Unconjugated | |
SUMF2 | |
2610040F05Rik; C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; DKFZp566I1024; DKFZp686I1024; DKFZp686L17160; DKFZp781L1035; FGE2; Inactive C-alpha-formylglycine-generating enzyme 2; MGC99485; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; PSEC0171; sulfatase modifying factor 2; sulfatase-modifying factor 2; Sumf2; UNQ1968/PRO4500 | |
Mouse | |
Protein A/G | |
RUO | |
25870 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction